EMAIL THIS PAGE TO A FRIEND
Expert opinion on pharmacotherapy
A review of the use of latanoprost for glaucoma since its launch.
PMID 22348427
Abstract
Prostaglandins are increasingly used as first choice treatment for glaucoma because they are highly effective, lack relevant systemic side effects and require just once-daily administration. Latanoprost is an ester prodrug analog of prostaglandin F2α, which reduces intraocular pressure (IOP) by increasing uveoscleral outflow. Latanoprost 0.005% has received European and US approval as the first-line drug for reducing IOP in patients with open-angle glaucoma or ocular hypertension. Following the recent patent expiry for Xalatan®, a number of latanoprost generics have entered the glaucoma market. This review, achieved through PubMed and Medline research methods, describes the composition, pharmacokinetics, mode of action, efficacy, side effects and safety profile of latanoprost. Latanoprost was the first prostaglandin analog introduced in glaucoma management and it dramatically changed the market of the disease thanks to its efficacy and safety. Conjunctival hyperemia, which is commonly found after latanoprost use, is associated with a minor efficacy and duration of trabeculectomy; yet, from the ophthalmologist's perspective, this side effect seems largely counterbalanced by the high efficacy and safety of this compound. It is always advisable to consider the pro-inflammatory mode of action of latanoprost because this may have negative effects in particular patients (i.e., those with uveitis and cystoid macular edema) for whom caution and close follow-up is necessary.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L1167
Latanoprost, ≥98% (HPLC), oil
C26H40O5