EMAIL THIS PAGE TO A FRIEND
Critical care (London, England)
Ethyl pyruvate for the treatment of acetaminophen intoxication: alternative to N-acetylcysteine?
PMID 22348679
Abstract
N-acetylcysteine is the classical antidote for acetaminophen overdose-induced hepatotoxicity, but its efficacy is limited by the need for early and only temporary treatment. Therefore, Yang and colleagues tested the hypothesis of whether ethyl pyruvate--another anti-inflammatory and antioxidant compound, which they had previously shown to protect against liver injury of various other etiologies--may allow circumventing these limitations. While ethyl pyruvate improved liver regeneration when administered early and during a limited period only, the opposite response was present both after delayed as well as prolonged treatment. The authors concluded that prolonged anti-inflammatory treatment is detrimental after acetaminophen intoxication-induced liver damage. On the one hand, this research paper confirms the need for biomarkers to monitor organ recovery after acetaminophen. On the other hand, this paper adds to the ongoing discussion on the usefulness of ethyl pyruvate as a resuscitation fluid in the critically ill.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W245712
Ethyl pyruvate, natural, FG
C5H8O3
E47808
Ethyl pyruvate, 98%
C5H8O3