EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Expedient synthesis of α-substituted fluoroethenes.
PMID 22349519
Abstract
A mild and efficient synthesis of 1-aryl-1-fluoroethenes from benzothiazolyl (aryl)fluoromethyl sulfones and paraformaldehyde, under DBU- or Cs(2)CO(3)-mediated conditions at room temperature, is described. A comparable diethyl fluoro(naphthalen-2-yl)methylphosphonate reagent does not react with paraformaldehyde under these mild conditions. The utility of the methodology for synthesis of terminal α-fluoroalkenes bearing electron-withdrawing functionalities is also shown.