EMAIL THIS PAGE TO A FRIEND
Amino acids
New approach for amino acid profiling in human plasma by selective fluorescence derivatization.
PMID 22349763
Abstract
A new approach for the separation of 6-aminoquinolyl-carbamyl (AQC)-derivatized amino acids has been proposed. The chromatography used ion-pairing mechanism to increase the method selectivity. Mobile phase was based on triethylamine buffer containing N,N-dimethyloctylamine as a modifier. A number of factors, buffer composition and pH, counterion concentration, temperature and acetonitrile gradient profile, were optimized to achieve final chromatographic conditions. With the presented analytical method, the separation and identification of 34 AQC-amino acids and amino compounds present in human plasma is possible. The results of validation proved the applicability of the method for quantification of 27 amino acids in biological samples. The ultrafiltration proposed as deproteinization procedure gave repeatable and reliable results for the amino acids under investigation. This method introduced in routine testing can be a suitable tool for amino acid profiling in plasma including all aspects of clinical application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90350
Triethylamine hydrochloride, ≥99.0% (AT)
C6H15N · HCl