EMAIL THIS PAGE TO A FRIEND
Molecular psychiatry
Attenuating the persistence of fear memory storage using a single dose of antidepressant.
PMID 22349784
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0115
MDL 11939, ≥98% (HPLC)
C20H25NO