EMAIL THIS PAGE TO A FRIEND
In vivo (Athens, Greece)
Black tea polyphenols inhibit tumor proteasome activity.
PMID 22351658
Abstract
Tea is a widely consumed beverage and its constituent polyphenols have been associated with potential health benefits. Although black tea polyphenols have been reported to possess potent anticancer activities, the effect of its polyphenols, theaflavins on the tumor's cellular proteasome function, an important biological target in cancer prevention, has not been carefully studied. Here black tea extract (T5550) enriched in theaflavins inhibited the chymotrypsin-like (CT) activity of the proteasome and proliferation of human multiple myeloma cells in a dose-dependent manner. Also an isolated theaflavin (TF-1) can bind to, and inhibit the purified 20S proteasome, accompanied by suppression of tumor cell proliferation, suggesting that the tumor proteasome is an important target whose inhibition is at least partially responsible for the anticancer effects of black tea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55016
Theaflavin, analytical standard
C29H24O12
53963 Theaflavin monogallate, analytical standard
C36H28O16