EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[ACE inhibitor and calcium antagonist in fixed combination: effective protection for hypertensive patients at risk].
PMID 22352259
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6668
Lercanidipine hydrochloride, ≥98% (HPLC)
C36H41N3O6 · HCl