EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Efficient recyclable CuI-nanoparticle-catalyzed S-arylation of thiols with aryl halides on water under mild conditions.
PMID 22354412
Abstract
CuI nanoparticles efficiently catalyzed the C-S cross coupling of aryl and alkyl thiols with aryl halides in the absence of ligands on water under mild conditions. A wide range of diaryl sulfides and aryl alkyl sulfides are synthesized in good to excellent yields utilizing this protocol. This procedure is particularly noteworthy given its mild conditions, avoiding the undesired formation of disulfides through oxidation of thiols. The recovery and successful reutilization of the catalyst is described. Furthermore, the directed synthesis of bisarylated product is presented.