EMAIL THIS PAGE TO A FRIEND
Nature
Structural biology: Muscarinic receptors become crystal clear.
PMID 22358836
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C002
(±)-Quinuclidinyl benzilate, powder
C21H23NO3