EMAIL THIS PAGE TO A FRIEND
The Lancet. Infectious diseases
Supply chain problems for Chagas disease treatment.
PMID 22361418
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3415
Nifurtimox, ≥98% (HPLC)
C10H13N3O5S