EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of a 4,8-dialkoxy-benzo[1,2-b:4,5-b']difuran unit and its application in photovoltaic polymer.
PMID 22361897
Abstract
A new building block of benzo[1,2-b:4,5-b']difuran (BDF) was firstly designed and synthesized. The newly designed unit was applied for constructing a new photovoltaic low band gap polymer, PBDFDTBT, which exhibited promising power conversion efficiency of 5.0%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
249416
2-Ethylhexyl bromide, contains 1% potassium carbonate as stabilizer, 95%
C8H17Br