EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Stereoselective synthesis of hydroxy stilbenoids and styrenes by atom-efficient olefination with thiophthalides.
PMID 22362491
Abstract
The synthesis of stilbenoids and styryl carboxylic acids is accomplished with high E-stereoselectivity by olefination of aldehydes with thiophthalides under basic conditions. The olefination is highly atom-efficient as it only loses elemental sulfur during the reaction. This olefination, in conjunction with retro Kolbe-Schmitt reaction, allows facile synthesis of E-hydroxystilbenoids with minimal employment of protecting groups. This study also discloses two important findings: formation of i) 4-methylsulfanyl isocoumarins and ii) an 2-arylindenone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P39605
Phthalide, 98%
C8H6O2
W419501
Phthalide, 98%
C8H6O2