EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology
[Enterofuril eradication therapy in chronic inflammatory diseases of the upper digestive tract].
PMID 22364005
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46494
Nifuroxazide, VETRANAL®, analytical standard
C12H9N3O5