EMAIL THIS PAGE TO A FRIEND
Journal of the European Academy of Dermatology and Venereology : JEADV
Treatment of livedoid vasculopathy with alprostadil (PGE-1): case report and review of published literature.
PMID 22364635