EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Efficient synthesis and biological evaluation of proximicins A, B and C.
PMID 22364744
Abstract
A quick and efficient synthesis and the biological evaluation of promising antitumor-antibiotics proximicins A, B and C are reported. The characteristic repetitive unit of these molecules, the methyl 4-Boc-aminofuran-2-carboxylate 15, was prepared in three synthetic steps in good yield using an optimised copper-catalysed amidation method. The proximicins were evaluated for their antitumor activity using cellular methods. Proximicin B induced apoptosis in both Hodgkin's lymphoma and T-cell leukemia cell lines and proximicin C exhibited significantly high cytotoxicity against glioblastoma and breast carcinoma cells. The proximicins were also screened against Escherichia coli, Enterococcus faecalis and several strains of methicillin-and multidrug-resistant Staphylococcus aureus. Proximicin B showed noteworthy activity against antibiotic-resistant Gram-positive cocci.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9653
Netropsin dihydrochloride, from Streptomyces netropsis, ≥98% (HPLC and TLC), powder
C18H26N10O3 · 2HCl