EMAIL THIS PAGE TO A FRIEND
Biomedical and environmental sciences : BES
Correlation of seven biological factors (Hsp90a, p53, MDM2, Bcl-2, Bax, Cytochrome C, and Cleaved caspase3) with clinical outcomes of ALK+ anaplastic large-cell lymphoma.
PMID 22365399
Abstract
To explore correlation of seven apoptosis-related proteins (Hsp90a, p53, MDM2, Bcl-2, Bax, Cytochrome C, and Cleaved caspase3) with clinical outcomes of ALK+ anaplastic large-cell lymphoma (ALCL). Using immunohistochemistry and immunofluorescence double staining methods, the expressions of these seven apoptosis-associated proteins were studied to clarify their relationship with clinical outcomes of 36 ALK+ and 25 ALK-systemic ALCL patients enrolled between 1996 and 2006. The relationship of these apoptosis-regulating proteins with NPM-ALK status was also evaluated with the tyrosine inhibitor herbimycin A (HA) in vitro by immunocytochemistry, Western blotting and flow cytometric assays. The presence of Hsp90α-, MDM2-, Bax-, Cytochrome C, and Cleaved caspase3-positive tumor cells was found significantly different in ALK+ and ALK-ALCLs, which was correlated with highly favorable clinical outcome. The Bcl-2- and p53-positive tumor cells were found in groups of patients with unfavorable prognosis. Inhibition of NPM-ALK by HA could reactivate the p53 protein and subsequent apoptosis-related proteins and therefore induced apoptosis in ALK+ ALCL cells. Our results suggest that these seven proteins might be involved in apoptosis regulation and associated with clinical outcome of ALK+ systemic ALCLs. We also reveal a dynamic chain relation that NPM-ALK regulates p53 expression and subsequent apoptosis cascade in ALK+ ALCLs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6649
Herbimycin A from Streptomyces hygroscopicus, ≥95% (HPLC), powder
C30H42N2O9