EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
Different effects of the selective serotonin reuptake inhibitors fluvoxamine, paroxetine, and sertraline on the pharmacokinetics of fexofenadine in healthy volunteers.
PMID 22367658
Abstract
Although the interaction between selective serotonin reuptake inhibitors (SSRIs) and other drugs is important in the treatment of depression, there have been few studies of SSRIs concerning transporter-mediated interactions in humans. The objective of this study was to evaluate the in vivo effects of commonly used SSRIs on the pharmacokinetics of fexofenadine, a P-glycoprotein substrate.Twelve healthy volunteers (3 females and 9 males) were enrolled in this study. Each subject received a 60-mg dose of fexofenadine orally at baseline. Afterward, they were randomly assigned to receive 3 treatments with a 60-mg dose of fexofenadine after a 7-day treatment with fluvoxamine (50 mg/d), paroxetine (20 mg/d), or sertraline (50 mg/d), with 2-week intervals between the agents.Fluvoxamine pretreatment significantly increased the maximum plasma concentration, the area under the concentration time curves, and the 24-hour urinary fexofenadine excretion by 66% (P = 0.004), 78% (P = 0.029), and 78% (P < 0.001), respectively, without prolonging its elimination half-life. Paroxetine extended the elimination half-life of fexofenadine by 45% (P = 0.042), and it increased the 24-hour urinary fexofenadine excretion by 55% (P = 0.002). Sertraline did not alter any of the pharmacokinetic parameters of fexofenadine.This is the first report of the different effects of 3 commonly used SSRIs on fexofenadine pharmacokinetics in humans. Our 7-day, repeated-dose clinical study in healthy volunteers indicates that fluvoxamine and paroxetine, but not sertraline, may impact the patient exposure to fexofenadine, which is likely the result of P-glycoprotein inhibition in the small intestine and/or the liver.