EMAIL THIS PAGE TO A FRIEND
Journal of applied genetics
Genetic variation of Fusarium oxysporum isolates forming fumonisin B(1) and moniliformin.
PMID 22367665
Abstract
Thirty single-spore isolates of a toxigenic fungus, Fusarium oxysporum, were isolated from asparagus spears and identified by species-specific polymerase chain reaction (PCR) and translation elongation factor 1-α (TEF) sequence analysis. In the examined sets of F. oxysporum isolates, the DNA sequences of mating type genes (MAT) were identified. The distribution of MAT idiomorph may suggest that MAT1-2 is a predominant mating type in the F. oxysporum population. F. oxysporum is mainly recognised as a producer of moniliformin-the highly toxic secondary metabolite. Moniliformin content was determined by high-performance liquid chromatography (HPLC) analysis in the range 0.05-1,007.47 μg g(-1) (mean 115.93 μg g(-1)) but, also, fumonisin B(1) was detected, in the concentration range 0.01-0.91 μg g(-1) (mean 0.19 μg g(-1)). There was no association between mating types and the mycotoxins biosynthesis level. Additionally, a significant intra-species genetic diversity was revealed and molecular markers associated with toxins biosynthesis were identified.