EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Follow-up of natural products isolation.
PMID 22367909
Abstract
Follow-up of natural products isolation refers to re-isolation of compound(s) of interest in larger amounts for further pharmacological testing, conclusive structure elucidation, structure modifications to synthesize analogs for structure-activity relationships (SAR) studies, preformulation and formulation studies or clinical trials. In addition to conventional synthetic chemistry approaches, several other methodologies can be applied for following-up natural products isolation. This chapter outlines, with specific examples, various strategies and methods involved in follow-up of natural products isolation.