EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Biosynthetic approach for functional protein microarrays.
PMID 22370272
Abstract
Protein microarrays have emerged as an indispensable research tool for providing information about protein functions and interactions through high-throughput screening. Traditional methods for immobilizing biomolecules onto solid surfaces have been based on covalent and noncovalent binding, entrapment in semipermeable membranes, microencapsulation, sol gel, and hydrogel methods. Each of these techniques has its own strengths but fails to combine the most important tenets of a functional protein microarray such as covalent attachment, native protein conformation, homogeneity of the protein monolayer, control over active site orientation, and retention of protein activity. Here we present a selective and site-directed covalent immobilization technique for proteins via a benzoxazine ring formation through a Diels-Alder reaction in water and a genetically encoded 3-amino-L-tyrosine (3-NH(2)Tyr) amino acid. Fully functional protein microarrays, with monolayer arrangements and complete control over their orientations, were generated using this strategy.