EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium-catalyzed intramolecular hydroarylation of 4-benzofuranyl alkynoates. Approach to angelicin derivatives.
PMID 22372387
Abstract
Intramolecular hydroarylation of 4-benzofuranyl alkynoates using Pd(OAc)(2) as catalyst took place selectively and efficiently, giving angular furocoumarin derivatives in high yields. The parent angelicin was obtained in 80% yield by this method. The starting 4-benzofuranyl alkynoates were easily accessible from readily available 4-hydroxybenzofurans and alkynoic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0956
Angelicin
C11H6O3