EMAIL THIS PAGE TO A FRIEND
Natural product research
Phytochemical composition of polar fraction of Stachys germanica L. subsp. salviifolia (Ten.) Gams, a typical plant of Majella National Park.
PMID 22372659
Abstract
In this study, we report the isolation and identification of several compounds present in the polar fraction of Stachys germanica L. subsp. salviifolia (Ten.) Gams, collected in the protected area of Majella National Park. In particular, we have isolated and identified harpagide, 7-β-hydroxy-harpagide, ajugol, 5-allosyloxy-aucubin, verbascoside and, for the first time in this genus, arbutin.