EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Ion size influence on the Ar solvation shells of M(+)-C6F6 clusters (M = Na, K, Rb, Cs).
PMID 22375689
Abstract
The size-specific influence of alkali metal ions in the gradual transition from cluster rearrangement to solvation dynamics is investigated by means of molecular dynamics simulations for alkali metal cation-hexafluorobenzene systems, M(+)-C(6)F(6) (M = Na, K, Rb and Cs), surrounded by Ar atoms. To analyze such transition, different small aggregates of the M(+)-C(6)F(6)-Ar(n) (n = 1, ..., 30) type and M(+)-C(6)F(6) clusters solvated by about 500 Ar atoms are considered. The Ar-C(6)F(6) interaction contribution has been described using two different formalisms, based on the interaction decomposition in atom-bond and in atom-effective atom terms, which have been applied to study the small aggregates and to investigate the Ar solvated M(+)-C(6)F(6) clusters, respectively. The selectivity of the promoted phenomena from the M(+) ion size and their dependence from the number of Ar atoms is characterized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326720
Hexafluorobenzene, ≥99.5%, NMR grade
C6F6
H8706
Hexafluorobenzene, 99%
C6F6