EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
New steroidal glycoside ester and aliphatic acid from the fruits of Lycium chinense.
PMID 22375866
Abstract
Two new compounds stigmast-5-en-3β-ol-3-O-β-D-(2'-n-triacontanoyl) glucopyranoside (1) and 19,21-dimethyl triacont-17,22,24,26,28-pentaene-1-oic acid (2), along with the three known compounds n-tetracosanyl octadec-9-enoate (3), β-sitosterol, and β-sitosterol-3-O-β-D-glucoside, have been isolated from the methanol extract of Lycium chinense fruits. The structures of these phytoconstituents have been established on the basis of spectral data analysis and chemical reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47132
Stigmasterol, certified reference material, 10 mg/mL in chloroform
C29H48O