EMAIL THIS PAGE TO A FRIEND
Restorative Neurology and Neuroscience
Third trimester amniotic fluid cells with the capacity to develop neural phenotypes and with heterogeneity among sub-populations.
PMID 22377907