EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Esterases with an introduced amidase-like hydrogen bond in the transition state have increased amidase specificity.
PMID 22378481