EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
High-performance broadband photodetector using solution-processible PbSe-TiO(2)-graphene hybrids.
PMID 22378495
Abstract
Highly sensitive, multicomponent broadband photodetector devices are made from PbSe/graphene/TiO(2). TiO(2) and PbSe nanoparticles act as light harvesting photoactive materials from the UV to IR regions of the electromagnetic spectrum, while the graphene acts as a charge collector for both photogenerated holes and electrons under an applied electric field.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254231
Lead(II) selenide, 99.99%
PbSe