EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Regioselective intramolecular [3+2] annulation of allene-nitrones.
PMID 22382420
Abstract
The regioselective intramolecular 1,3-dipolar cycloaddition of the phenylsulfonylallene-nitrone derivatives has been developed. This reaction showed that the distal double bond of the allene exclusively reacted with the nitrone group to produce the bicyclic isoxazolidine derivatives regardless of the substitution pattern on the allenyl moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
294985
Allene, ≥95%
C3H4