EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Molecular symmetry and biological activities of new symmetrical tris(2-aminoethyl)amine derivatives.
PMID 22382425
Abstract
In terms of molecular symmetry and bioactivity, new C3- and CS-symmetrical derivatives based on the tris(2-aminoethyl)amine scaffold were designed and synthesized. The synthesized compounds were evaluated for antiviral activity with herpes simplex virus type 1 (HSV-1) by a plaque reduction assay and for cytotoxic activity with Vero cells. Most of the compounds showed no significant anti-HSV-1 activity, but some of the symmetrical derivatives showed high levels of cytotoxic activitiy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
275484
trans-2,3-Dimethoxycinnamic acid, 97%
C11H12O4
225630
Tris(2-aminoethyl)amine, 96%
C6H18N4