EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
Physicochemical investigations and stability studies of amorphous gliclazide.
PMID 22382730
Abstract
Gliclazide (GLI), a poorly water-soluble antidiabetic, was transformed into a glassy state by melt quench technique in order to improve its physicochemical properties. Chemical stability of GLI during formation of glass was assessed by monitoring thin-layer chromatography, and an existence of amorphous form was confirmed by differential scanning calorimetry and X-ray powder diffractometry. The glass transition occurred at 67.5°C. The amorphous material thus generated was examined for its in vitro dissolution performance in phosphate buffer (pH 6.8). Surprisingly, amorphous GLI did not perform well and was unable to improve the dissolution characteristics compared to pure drug over entire period of dissolution studies. These unexpected results might be due to the formation of a cohesive supercooled liquid state and structural relaxation of amorphous form toward the supercooled liquid region which indicated functional inability of amorphous GLI from stability point of view. Hence, stabilization of amorphous GLI was attempted by elevation of T(g) via formation of solid dispersion systems involving comprehensive antiplasticizing as well as surface adsorption mechanisms. The binary and ternary amorphous dispersions prepared with polyvinylpyrrolidone K30 (as antiplasticizer for elevation of T (g)) and Aerosil 200® and/or Sylysia® 350 (as adsorbent) in the ratio of 1:1:1 (w/w) using kneading and spray-drying techniques demonstrated significant enhancement in rate and extent of dissolution of drug initially. During accelerated stability studies, ternary systems showed no significant reduction in drug dissolution performance over a period of 3 months indicating excellent stabilization of amorphous GLI.