PloS one
In vitro study of novel collagenase (XIAFLEX®) on Dupuytren's disease fibroblasts displays unique drug related properties.
PMID 22384021
Abstract
Dupuytren's disease (DD) is a benign, fibroproliferative disease of the palmar fascia, with excessive extracellular matrix (ECM) deposition and over-production of cytokines and growth factors, resulting in digital fixed flexion contractures limiting hand function and patient quality of life. Surgical fasciectomy is the gold standard treatment but is invasive and has associated morbidity without limiting disease recurrence. Injectable Collagenase Clostridium histolyticum (CCH)--Xiaflex®--is a novel, nonsurgical option with clinically proven in vivo reduction of DD contractures but with limited in vitro data demonstrating its cellular and molecular effects. The aim of this study was to delineate the effects of CCH on primary fibroblasts isolated from DD and non-DD anatomical sites (using RTCA, LDH, WST-1, FACS, qRT-PCR, ELISA and In-Cell Quantitative Western Blotting) to compare the efficacy of varying concentrations of Xiaflex® against a reagent grade Collagenase, Collagenase A. Results demonstrated that DD nodule and cord fibroblasts had greater proliferation than those from fat and skin. Xiaflex® exposure resulted in dose- and time-dependent inhibition of cellular spreading, attachment and proliferation, with cellular recovery after enzyme removal. Unlike Collagenase A, Xiaflex® did not cause apoptosis. Collagen expression patterns were significantly (p<0.05) different in DD fibroblasts across anatomical sites - the highest levels of collagen I and III were detected in DD nodule, with DD cord and fat fibroblasts demonstrating a smaller increase in both collagen expression relative to DD skin. Xiaflex® significantly (p<0.05) down-regulated ECM components, cytokines and growth factors in a dose-dependent manner. An in vitro scratch wound assay model demonstrated that, at low concentrations, Xiaflex® enabled a faster fibroblast reparatory migration into the wound, whereas, at high concentrations, this process was significantly (p<0.05) inhibited. This is the first report elucidating potential mechanisms of action of Xiaflex® on Dupuytren fibroblasts, offering a greater insight and a better understanding of its effect in DD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1639
Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9891
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
C5894
Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C2674
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
C9722
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, cell culture tested
C1764
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid
C1889
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C2139
Collagenase from Clostridium histolyticum, release of rat epididymal adipocytes and hepatocytes tested (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9263
Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
C2014
Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
C7657
Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
C9407
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C9697
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C0255
Collagenase from Clostridium histolyticum, purified by chromatography, ≥1 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
C0773
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, 1,000-3,000 CDU/mg solid (CDU = collagen digestion units)
C2399
Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, 1,000-3,000 CDU/mg solid
C2799
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, 1000-3000 CDU/mg solid (CDU = collagen digestion units), ≥4 FALGPA units/mg solid, high purity
C8051
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
C8176
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
C7926
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
C0374
Collagenase from Clostridium histolyticum, Animal component-free, ≥300 CDU/mg solid