EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Cavitation-induced radical-chain oxidation of valeric aldehyde.
PMID 22386946
Abstract
The application of high-amplitude ultrasound to liquids triggers cavitation. By the collapse of the thereby appearing vacuum cavities, high temperatures can be reached in a transient manner. The high temperatures in these hot-spots can lead to homolytic scission of chemical bonds. The thereby generated radicals are usually utilized in aqueous systems for the degeneration of organic pollutants. In this contribution, we demonstrate that the radicals can also be used for synthetic purposes: under an oxygen atmosphere, they trigger the oxidation of an aldehyde substrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
110132
Valeraldehyde, 97%
C5H10O
W309818
Valeraldehyde, ≥97%, FG
C5H10O