EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Enantioselective synthesis of tryptophan derivatives by a tandem Friedel-Crafts conjugate addition/asymmetric protonation reaction.
PMID 22390403
Abstract
The tandem Friedel-Crafts conjugate addition/asymmetric protonation reaction between 2-substituted indoles and methyl 2-acetamidoacrylate is reported. The reaction is catalyzed by (R)-3,3'-dibromo-BINOL in the presence of stoichiometric SnCl(4), and is the first example of a tandem conjugate addition/asymmetric protonation reaction using a BINOL·SnCl(4) complex as the catalyst. A range of indoles furnished synthetic tryptophan derivatives in good yields and high levels of enantioselectivity, even on a preparative scale. The convergent nature of this transformation should lend itself to the preparation of unnatural tryptophan derivatives for use in a broad array of synthetic and biological applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317519
Methyl 2-acetamidoacrylate, 98%
C6H9NO3