EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Suzuki coupling of potassium cyclopropyl- and alkoxymethyltrifluoroborates with benzyl chlorides.
PMID 22390789
Abstract
Efficient Csp(3)-Csp(3) Suzuki couplings have been developed with both potassium cyclopropyl- and alkoxymethyltrifluoroborates. Moderate to good yields have been achieved in the cross-coupling of potassium cyclopropyltrifluoroborate with benzyl chlorides possessing electron-donating or electron-withdrawing substituents. Benzyl chloride was also successfully cross-coupled to potassium alkoxymethyltrifluoroborates derived from primary, secondary, and tertiary alcohols.