EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Targeting Werner syndrome protein sensitizes U-2 OS osteosarcoma cells to selenium-induced DNA damage response and necrotic death.
PMID 22390926
Abstract
Mutations in the Werner syndrome protein (WRN), a caretaker of the genome, result in Werner syndrome, which is characterized by premature aging phenotypes and cancer predisposition. Methylseleninic acid (MSeA) can activate DNA damage responses and is a superior compound to suppress tumorigenesis in mouse models of cancer. To test the hypothesis that targeting WRN can potentiate selenium toxicity in cancer cells, isogenic WRN small hairpin RNA (shRNA) and control shRNA U-2 OS osteosarcoma cells were treated with MSeA for 2d, followed by recovery for up to 7d. WRN deficiency sensitized U-2 OS cells to MSeA-induced necrotic death. Co-treatment with the ataxia-telangiectasia mutated (ATM) kinase inhibitor KU55933 desensitized the control shRNA cells, but not WRN shRNA cells, to MSeA treatment. WRN did not affect MSeA-induced ATM phosphorylation on Ser-1981 or H2A.X phosphorylation on Ser-139, but promoted recovery from the MSeA-induced DNA damage. Taken together, WRN protects U-2 OS osteosarcoma cells against MSeA-induced cytotoxicity, suggesting that oxidative DNA repair pathway is a promising target for improving the efficacy of selenium on tumor suppression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
541281
Methaneseleninic acid, 95%
CH4O2Se