EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of tropane alkaloids atropine and scopolamine by liquid chromatography-mass spectrometry in plant organs of Datura species.
PMID 22391493
Abstract
Hyoscyamine (atropine) and scopolamine are the predominant tropane alkaloids in the Datura genus, occurring in all plant organs. The assessment of the alkaloid content of various plant parts is essential from the viewpoint of medical use, but also as a potential risk of toxicity for humans and animals. Therefore, a reliable method for the determination of tropane alkaloid content is of high importance. The present work aimed at the elaboration of a rapid method for determination of the most abundant Datura alkaloids by LC-MS technique using a new generation of core-shell particle packed column. Tropane alkaloid content was investigated in various plant organs of four Datura taxa (D. innoxia, D. metel, D. stramonium, and D. stramonium var. tatula), grown under the same conditions, in two developmental stages. We have developed a rapid LC-MS method for the quantitative determination of atropine and scopolamine, which was successfully applied to quantify the alkaloids in different plant organs (leaves, flowers, stems, seeds) of thorn apples after a simple sample preparation step. Elaboration and validation of the method and analysis of plant extracts were done by UFLC-MS technique, employing an Ascentis Express C18 column. Detection was done in positive ionization mode (ESI+) and the method suitability was evaluated by several validation characteristics. Quantitation limits are 333 and 167 pgmL(-1) for scopolamine and atropine, respectively, and the method shows very good repeatability. The analysis of Datura extracts revealed significant differences depending on the species, the organ and the sampling period. Atropine was found to be dominant over scopolamine in three out of the four taxa investigated. D. innoxia showed the highest concentrations of scopolamine in all organs examined, whereas D. metel accumulated the lowest scopolamine levels. Hyoscyamine, measured as atropine, was the highest in D. stramonium var. tatula, and the lowest in D. innoxia. Samples collected in summer had higher scopolamine levels than autumn samples, concerning both stems and leaves.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50461-U Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
50462-U Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
50822-U Ascentis® Express C18, 2 Micron Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
50805-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
50809-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm
50811-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
50812-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
50813-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
50814-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
50815-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
50816-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
50817-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 3 mm
50819-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
50821-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
53982-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 75 μm
53985-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 100 μm
53989-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 200 μm
53992-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
53998-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
54219-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 75 μm
54256-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 100 μm
54261-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 200 μm
54271-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
54273-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
53501-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
53504-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
53508-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
53799-U Ascentis® Express C18, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
53802-U Ascentis® Express C18, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
53804-U Ascentis® Express C18, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm