EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Histopathological effects of carbaryl on digestive system of snake-eyed lizard, Ophisops elegans.
PMID 22392006
Abstract
We examined the effects of carbaryl in the digestive system of Ophisops elegans. Lizards were exposed once to different concentrations of carbaryl (2.5, 25 and 250 μg/g). After 96 h, findings related to the esophagus in all treatment groups were not conspicuous. The most important histological defects were observed in the stomach. In the small intestine, collapse of villi was prominent at high-dose. In the large intestine, disintegration in epithelial cells and scattered secretory granules of goblet cells were observed at high dose.