EMAIL THIS PAGE TO A FRIEND
Anesthesia and analgesia
Cyanide toxicity in juvenile pigs and its reversal by a new prodrug, sulfanegen sodium.
PMID 22392971
Abstract
Cyanide (CN) toxicity is a serious clinical problem and can occur with sodium nitroprusside (SNP) administration, accidental smoke inhalation, industrial mishaps, and bio-terrorism. In this study, we induced severe CN toxicity independently with SNP or sodium cyanide (NaCN) in a juvenile pig model to demonstrate reversal of severe CN toxicity with a new antidote, sulfanegen sodium, a prodrug of 3-mercaptopyruvate. SNP study: A pilot study in 11 anesthetized, mechanically ventilated juvenile pigs allowed us to determine the dose of SNP to induce CN toxicity. Blood CN, serum lactates, and blood gases were monitored. CN toxicity was defined as the occurrence of severe lactic acidosis accompanied by significant elevation in blood CN levels. Based on this pilot study, 8 anesthetized pigs received a high-dose i.v. infusion of SNP (100 mg/h) for 2 hours to induce CN toxicity. They were then randomized to receive either sulfanegen sodium or placebo. Four pigs received 3 doses of sulfanegen sodium (2.5 g i.v.) every hour after induction of severe CN toxicity, and 4 pigs received placebo. NaCN study: A pilot study was conducted in 4 spontaneously ventilating pigs sedated with propofol plus ketamine to demonstrate hemodynamic and metabolic stability for several hours. After this, 6 pigs were similarly sedated and given NaCN in bolus aliquots to produce CN toxicity ultimately resulting in death. Hemodynamics and metabolic variables were followed to define peak CN toxicity. In another group of 6 pigs, severe CN toxicity was induced by this method, and at peak toxicity, the animals were given sulfanegen sodium (2.5 g i.v.) followed by a repeat dose 60 minutes later in surviving animals. SNP study: The pilot study demonstrated the occurrence of a significant increase in blood CN levels (P < 0.05) accompanied by severe lactic acidemia (P < 0.05) in all pigs receiving a high dose of SNP. Administration of the sulfanegen antidote resulted in progressive significant reduction in blood lactate and CN levels with 100% survival (P < 0.05), whereas the placebo-treated pigs deteriorated and did not survive (P < 0.05). NaCN study: NaCN injection resulted in CN toxicity accompanied by severe lactic acidosis and mortality in all the pigs. Sulfanegen sodium reversed this toxicity and prevented mortality in all the pigs treated with this antidote. CN toxicity can be successfully induced in a juvenile pig model with SNP or NaCN. The prodrug, sulfanegen sodium, is effective in reversing CN toxicity induced by SNP or NaCN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359394
Sodium mercaptopyruvate, ≥85%, technical grade
C3H3NaO3S