EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Rapid determination of 6 pesticide residues in tomato paste by ultra performance liquid chromatography-tandem mass spectrometry].
PMID 22393693
Abstract
An ultra performance liquid chromatography-tandem mass spectrometric method (UPLC-MS/MS) was established for the simultaneous determination of imidacloprid, carbendazim, thiophanate-methyl, propamocarb, methomyl and dimethomorph residues in tomato paste. The samples were extracted by methanol-water (1: 1, v/v) containing 0.1% (v/v) acetic acid. The separation was performed on a Waters Acquity UPLC system with a BEH C18 column with the gradient elution of methanol and water (containing 10 mmol/L ammonium acetate). The six pesticides were determined in the modes of electrospray positive ionization (ESI+) and multiple reaction monitoring (MRM). The analytes were quantified by matrix-matched standard solution, and the calibration curves showed good linearity within the concentrations of 0.005 to 0.2 mg/L and the correlation coefficients (r) were more than 0.995. The average recoveries of the six pesticides ranged from 66.8% to 102.9% in the three spiked levels of 0.02, 0.05 and 0.2 mg/kg. The relative standard deviations (RSDs) were all less than 15%. The limits of quantification (LOQ, S/N > 10) were 0.02 mg/kg for the all analytes. The results indicate that the method is easier, faster, more sensitive, and suitable for the qualitative and quantitative confirmation of the six pesticide residues from tomato paste.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45688
Thiophanate-methyl, PESTANAL®, analytical standard
C12H14N4O4S2