EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
[Ultrastructural cellular alterations of the natronophilic endoevaporite cyanobacterium Euhalothece natronophila during fossilization].
PMID 22393763
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1362
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H6CaO4