EMAIL THIS PAGE TO A FRIEND
Organic letters
Palladium-catalyzed N-arylation of 2-aminothiazoles.
PMID 22394197
Abstract
A method for the Pd-catalyzed coupling of 2-aminothiazole derivatives with aryl bromides and triflates is described. Significantly, for this class of nucleophiles, the coupling exhibits a broad substrate scope and proceeds with a reasonable catalyst loading. Furthermore, an interesting effect of acetic acid as an additive is uncovered that facilitates catalyst activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123129
2-Aminothiazole, 97%
C3H4N2S