EMAIL THIS PAGE TO A FRIEND
Mycopathologia
Evaluation of pathological effects in broilers during fumonisins and clays exposure.
PMID 22396010
Abstract
This study was conducted to evaluate the possible protector effect of bentonite and zeolite in Bovans chicks fed a diet containing 59 mg kg(-1) of fumonisin B1 (FB1) during 3 weeks. A total of 200 one-day-old male chicks were treated varying the amount of bentonite and zeolite. Chick weight was registered weekly. At the end of the experiment, all the chicks were killed, and the livers were analyzed for gross examination and histopathological changes. Plasmatic activity of alanine amino transferase and aspartate amino transferase (AST) were also determined. Sphinganine and the sphinganine-to-sphingosine ratio in serum were evaluated. Both, bentonite and zeolite showed a protector effect against FB1 adsorption in the digestive tract of chicks. Chicks fed with FB1-contaminated feed, amended either with zeolite or bentonite, were heavier, and no macroscopic lesions were observed in the livers. AST activity might be considered as an indicator for FB1 exposition because AST levels were affected when only FB1 was present in the basal diet. These results indicate that both, zeolite and bentonite can be added into feed to diminish the effects of FB1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4556
D-threo-Dihydrosphingosine
C18H39NO2
D4681
L-threo-Dihydrosphingosine, ≥95% (TLC)
C18H39NO2