EMAIL THIS PAGE TO A FRIEND
The British journal of surgery
Effects of tirapazamine on experimental colorectal liver metastases after radiofrequency ablation (Br J Surg 2012; 99: 567-575).
PMID 22396055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0552
Tirapazamine, ≥98% (HPLC)
C7H6N4O2