EMAIL THIS PAGE TO A FRIEND
Nihon Shokakibyo Gakkai zasshi = The Japanese journal of gastro-enterology
[Recent approaches of personalized medicine for inflammatory bowel disease].
PMID 22398899
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T27502
L-4-Thiazolidinecarboxylic acid, 98%
C4H7NO2S