EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Xanthones in mangosteen juice are absorbed and partially conjugated by healthy adults.
PMID 22399525
Abstract
The proposed health-promoting effects of the pericarp from mangosteen fruit have been attributed to a family of polyphenols referred to as xanthones. The purpose of this study was to determine the bioavailability of xanthones from 100% mangosteen juice in healthy adult participants (n = 10). Pericarp particles accounted for 1% of the mass and 99% of the xanthone concentration in the juice. The juice provided 5.3 ± 0.1 mmol/L total xanthones with α-mangostin, garcinones (C, D, and E), γ-mangostin, gartanins, and other identified xanthones accounting for 58, 2, 6, 4, and 5%, respectively. Participants ingested 60 mL mangosteen juice with a high-fat breakfast. Free and conjugated (glucuronidated/sulfated) xanthones were detected in serum and urine. There was marked variation in the AUC (762-4030 nmol/L × h), maximum concentration (113 ± 107 nmol/L), and time to maximum concentration (3.7 ± 2.4 h) for α-mangostin in sera during the 24-h collection. Similarly, xanthones in 24-h urine ranged from 0.9 to 11.1 μmol and accounted for 2.0 ± 0.3% (range 0.3-3.4%) of the ingested dose. There were no significant differences between female and male participants in mean pharmacokinetic values of α-mangostin in serum and urinary xanthones. Only 15.4 ± 0.7% of total xanthones in pericarp particles in the juice partitioned into mixed micelles during in vitro digestion. These results show that xanthones in mangosteen juice are absorbed when ingested along with a high-fat meal, although release of xanthones from pericarp particles during digestion may be limited.