EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Palladium-catalyzed S-benzylation of unprotected mercaptobenzoic acid with benzyl alcohols in water.
PMID 22402746
Abstract
Palladium-catalyzed S-benzylation of unprotected mercaptobenzoic acids with benzyl alcohols gave only S-benzylated mercaptobenzoic acids in good yields. Water may play an important role for the smooth generation of the (η(3)-benzyl)palladium species by activation of the hydroxyl group of the benzyl alcohol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S