EMAIL THIS PAGE TO A FRIEND
American journal of respiratory cell and molecular biology
A novel prostacyclin agonist protects against airway hyperresponsiveness and remodeling in mice.
PMID 22403804
Abstract
Airway remodeling in bronchial asthma results from chronic, persistent airway inflammation. The effects of the reversal of airway remodeling by drug interventions remain to be elucidated. We investigated the effects of ONO-1301, a novel prostacyclin agonist with thromboxane inhibitory activity, on the prevention and reversibility of airway remodeling in an experimental chronic asthma model. Mice sensitized and challenged to ovalbumin (OVA) three times a week for 5 consecutive weeks were administered ONO-1301 or vehicle twice a day from the fourth week of OVA challenges. Twenty-four hours after the final OVA challenge, airway hyperresponsiveness (AHR) was assessed, and bronchoalveolar lavage was performed. Lung specimens were excised for staining to detect goblet-cell metaplasia, airway smooth muscle, and submucosal fibrosis. Mice administered ONO-1301 showed limited increases in AHR compared with mice administered the vehicle. The histological findings of airway remodeling were improved in ONO-1301-treated mice compared with vehicle-treated mice. Presumably, these therapeutic effects of ONO-1301 are attributable to the up-regulation of production of hepatocyte growth factor (HGF) in lung tissue, because the neutralization of HGF by antibodies prevented the effects of ONO-1301 on AHR and airway remodeling. Mice administered ONO-1301 showed similar levels of AHR and airway remodeling as mice administered montelukast, a cysteinyl-leukotriene-1 receptor antagonist, and lower levels were observed in mice administered dexamethasone. These data suggest that ONO-1301 exerts the effect of reversing airway remodeling, at least in part through an elevation of HGF in the lungs, and may be effective as an anti-remodeling drug in the treatment of asthma.