EMAIL THIS PAGE TO A FRIEND
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi
[Research on molecular mechanism of drug resistance of adefovir dipivoxil].
PMID 22404035
Abstract
Adefovir dipivoxil (ADV) is a nucleotide anti-hepatitis B virus drug, which is able to effectively repress the replication of hepatitis B virus (HBV). ADV is a reverse transcriptase inhibitor. However, with the prolonged period of using, many kinds of HBV genetic mutants will occur and therefore resistance to ADV will be created. The purpose of this review is to draw the attention to the molecular mechanism of resistance to ADV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9730
Adefovir dipivoxil
C20H32N5O8P