EMAIL THIS PAGE TO A FRIEND
Natural product research
Pharmacokinetic study of arctigenin in rat plasma and organ tissue by RP-HPLC method.
PMID 22404522
Abstract
A high-performance liquid chromatography (HPLC) technique was developed for the determination of arctigenin in plasma and various organs of rats after the oral administration of 30, 50 and 70 mgkg(-1) of arctigenin to the Sprague-Dawley rats. Results showed that the validated HPLC method was simple, fast, reproducible and suitable to the determination of arctigenin in rat plasma and organ tissue and one-compartmental model with zero-order absorption process can well describe the changes of arctigenin concentration in the plasma. The concentration of compound was highest in the spleen, less in the liver and the least in the lung.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00075
(−)-Arctigenin, ≥95% (LC/MS-ELSD)
C21H24O6