EMAIL THIS PAGE TO A FRIEND
Biotechnology journal
Biosafety evaluation of recombinant protein production in goat mammary gland using adenoviral vectors: preliminary study.
PMID 22407796
Abstract
The production of recombinant proteins in the milk of non-transgenic goats can be achieved by transducing the mammary gland with recombinant adenoviral vectors. However, this process involves several regulatory issues. The current study evaluates the biosafety of this production system. We present a preliminary biosafety profile based on detection of adenoviral particles in different body fluids and the antibody response after adenoviral transduction of the goat mammary gland. In addition, two methods of adenoviral inactivation in milk were tested. Although adenoviral particles were detected in the milk until day 4 after transduction, they were absent in serum, saliva, urine and feces. Anti-adenovirus antibodies were detected in serum and milk. The virus inactivation methods neutralized adenoviral particles and preserved the immunological identity of the recombinant protein. These results support the idea of a safe production of recombinant proteins using adenoviral vectors.